Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium cyclopentadienide 2-3M in THF

Supplier: STREM CHEMICALS INC MS
Danger

93-1075-100MMOL 93-1075-500MMOL
100197-660EA 334.03 USD InStock
100197-660 100197-662
Sodium cyclopentadienide 2-3M in THF
Sodium cyclopentadienide
Metallocenes, Derivatives & Cp Precursors


Formula: C₅H₅Na
Density: 0,939
Flash Pt: 1 °F
MDL Number: MFCD00012161
CAS Number: 4984-82-1
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty