Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(diphenyl)silane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: DichlorodiphenylsilaneDiphenyldichlorosilane

93-1407-250G 93-1407-50G
100206-938EA 203.82 USD InStock
100206-938 100197-698
Dichloro(diphenyl)silane ≥97%
Dichloro(diphenyl)silane


Formula: C₁₂H₁₀Cl₂Si
MW: 253.2 g/mol
Boiling Pt: 304 °C
Melting Pt: –22 °C
Density: 1.204
Flash Pt: 316 °F
MDL Number: MFCD00000489
CAS Number: 80-10-4
UN: 1769
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty