Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(methyl)(phenyl)silane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Methylphenyldichlorosilane

93-1415-500G 93-1415-100G
100197-198EA 93.42 USD InStock
100197-198 100197-200
Dichloro(methyl)(phenyl)silane ≥97%
Dichloro(methyl)(phenyl)silane


Formula: C₇H₈Cl₂Si
MW: 191.13 g/mol
Boiling Pt: 205 °C
Density: 1.187
Flash Pt: 181 °F
MDL Number: MFCD00000490
CAS Number: 149-74-6
UN: 2437
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty