Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) phosphate hydrate

Supplier: STREM CHEMICALS INC MS

93-2713-50G 93-2713-250G
100210-312EA 128.8 USD InStock
100210-312 100210-310
Cobalt(II) phosphate hydrate
Cobalt(II) phosphate hydrate


Formula: Co₃(PO₄)₂·xH₂O
Density: 2,769
MDL Number: MFCD00150916
CAS Number: 10294-50-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty