Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) oxalate dihydrate

Supplier: STREM CHEMICALS INC MS
Synonyms: Nickel oxalate dihydrate

93-2831-50G 93-2831-250G
100211-246EA 196.74 USD InStock
100211-246 100207-214
Nickel(II) oxalate dihydrate
Nickel(II) oxalate dihydrate


Formula: C₂H₄NiO₆
MW: 182.741 g/mol
MDL Number: MFCD00167155
CAS Number: 6018-94-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty