Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) gluconate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Copper(II) D-gluconateCopper GluconateCopper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoateGluconic Acid Copper(II) Salt

93-2927-500G 93-2927-100G
100200-550EA 179.75 USD InStock
100200-550 100196-596
Copper(II) gluconate ≥98%
Copper(II) gluconate


Formula: C₁₂H₂₂CuO₁₄
MW: 453.84 g/mol
MDL Number: MFCD00075297
CAS Number: 13005-35-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty