Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc naphthenate 65% in mineral spirits (10% Zn)

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Naphthenic Acid Zinc Salt

93-3037-2000G 93-3037-500G
100204-304EA 103.32 USD InStock
100204-304 100210-350
Zinc naphthenate 65% in mineral spirits (10% Zn)
Zinc naphthenate


Formula: 2(C₁₁H₇O₂)·Zn
MW: 319.71 g/mol
Density: 1,01
Flash Pt: >100 °F
MDL Number: MFCD00147699
CAS Number: 12001-85-3
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty