Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutyltin dilaurate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Di-n-butyltin dilaurate

93-5009-250G 93-5009-1000G
100196-702EA 210.89 USD InStock
100196-702 100207-854
Dibutyltin dilaurate ≥98%
Dibutyltin dilaurate


Formula: C₃₂H₆₄O₄Sn
MW: 631.57 g/mol
Melting Pt: 24 °C
Density: 1.066
Flash Pt: >230 °F
MDL Number: MFCD00008963
CAS Number: 77-58-7
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty