Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(IV) oxide, hydrated ≥85%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Cerium(IV) hydroxide

93-5810-100G 93-5810-25G
100208-200EA 226.46 USD InStock
100208-200 100202-218
Cerium(IV) oxide, hydrated ≥85%
Cerium(IV) oxide, hydrated


Formula: CeO₂.xH₂O
MW: 208.15 g/mol
MDL Number: MFCD00010927
CAS Number: 12014-56-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty