Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(III) phosphate ≥99.9% (Ce basis)

Supplier: STREM CHEMICALS INC MS

93-5818-25G 93-5818-100G
100205-686EA 111.82 USD InStock
100205-686 100210-498
Cerium(III) phosphate ≥99.9% (Ce basis)
Cerium(III) phosphate


Formula: CeO₄P
MW: 235.09 g/mol
MDL Number: MFCD00049424
CAS Number: 13454-71-2
EINECS: 721880-19-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty