Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Europium(III) oxalate hydrate ≥99.9% (Eu basis)

Supplier: STREM CHEMICALS INC MS

93-6325-2G 93-6325-10G
100202-248EA 72.18 USD InStock
100202-248 100209-174
Europium(III) oxalate hydrate ≥99.9% (Eu basis)
Europium(III) oxalate hydrate


Formula: C₆Eu₂O₁₂·xH₂O
MW: 567.98 g/mol& (anhy)
MDL Number: MFCD00150143
CAS Number: 14175-02-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty