Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Europium(III) sulfate octahydrate ≥99.99% (Eu basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-6330-5G 93-6330-1G
100200-188EA 69.35 USD InStock
100200-188 100208-216
Europium(III) sulfate octahydrate ≥99.99% (Eu basis), PURATREM
Europium (III) sulphate octahydrate


Formula: Eu₂O₁₂S₃·8H₂O
MW: 736.23 g/mol
MDL Number: MFCD00149703
CAS Number: 10031-55-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty