Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dihydrogen hexahydroxyplatinate(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Hydrogen hexahydroxyplatinate(IV)

93-7806-5G 93-7806-1G
100198-458EA 1068.62 USD InStock
100198-458 100198-456
Dihydrogen hexahydroxyplatinate(IV) ≥99%
Dihydrogen hexahydroxyplatinate(IV)


Formula: H₂Pt(OH)₆
MW: 299.14 g/mol
MDL Number: MFCD00058744
CAS Number: 51850-20-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty