Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Diaminocyclohexanetetraacetic acid monohydrate ≥98.5% for complexometry, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: DCTA

34588-25G 34588-100G
BJ34588-25GEA 257.06 USD InStock
BJ34588-25G BJ-34588-100G
1,2-Diaminocyclohexanetetraacetic acid monohydrate ≥98.5% for complexometry, Fluka™
1,2-Diaminocyclohexanetetraacetic acid monohydrate


Formula: C₁₄H₂₂N₂O₈·H₂O
MW: 364.35 g/mol
Melting Pt: 210-220 °C
Storage Temperature: Ambient
MDL Number: MFCD00150952
CAS Number: 145819-99-4
Order Now
Specification Test Results
Assay (calculated on the anhydrous basis) Min. 99 %
Water (Karl Fischer) 4.8 - 5.6 %
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Sulfate (SO₄) Max. 0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty