Would you like to log in again?
Drum Wrench, Justrite®
Supplier: Justrite
08805
76006-510EA 120.99 USD InStock
76006-510
Drum Wrench, Justrite®
Wrenches
Drum Wrench opens drum bung caps.
  • Rugged and non-sparking
  • Design of handle protects the hand
This corrosive-resistant tool fits many 2" or 3/4" bung styles commonly used on drums.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty