Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formic acid ≥99.5%

Supplier: 0000036298
Synonyms: Methanoic acid

A117-50
MSPP-A117-50EA 93.85 USD InStock
MSPP-A117-50
Formic acid ≥99.5%
Formic acid


Formula: HCO₂H
MW: 46.03 g/mol
MDL Number: MFCD00003297
CAS Number: 64-18-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/23/723723.jpg
Order Now
Specification Test Results
Purity 99.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty