Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: HaliteSea saltSaline

746398-6X500G 746398-4X2.5KG
BJ746398-500GCS 346.14 USD InStock
BJ746398-500G BJ746398-2.5KG
Sodium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
Sodium chloride
Free flowing salt.


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1.465 °C (1.013 hPa)
Melting Pt: 800 °C
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now
Specification Test Results
Assay 99.0 - 101.5 %
Turbidity of solution complying
Colour of the solution complying
Flowability complying
pH (5 %, 25 °C) 5.0 - 9.0
Water insoluble matter Max. 0.005 %
Barium (Ba) complying
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.001 %
Heavy metals (ICP) ACS Max. 5 ppm
Bromide (Br) Max. 0.01 %
Chlorate, Nitrate (as NO₃) Max. 0.003 %
Iodide (I) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.004 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty