Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride)

Supplier: G-Biosciences
Synonyms: 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride

786-053 786-053
82022-390EA 1150.45 USD InStock
CA82022-390 82022-390
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)


Formula: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
MDL Number: MFCD00132962
CAS Number: 30827-99-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty