Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Microscopy Glass Slide, 12 Well Chamber, Removable, Ibidi USA MS
Supplier: IBIDI USA MS
81201
MSPP-81201PK 292.31 USD InStock
MSPP-81201
Microscopy Glass Slide, 12 Well Chamber, Removable, Ibidi USA MS
Slides Chamber Slides
Chamber slide with a removable well silicone chamber for cell culture and immunofluorescence staining.
  • Long-term sample storage
  • Self-adhesive silicone gasket mounted on a standard glass slide
  • Compatible with all common mounting media
Suitable for use with upright and inverted microscopes. Small staining volume for reduction of required medium, cells, and supplements. The ideal chambered slide for immunofluorescence assays and long-term sample storage. Non-fluorescent glass slide with frosted ends for easy handling and labeling.
Bottom: Glass slide (26×76×1 mm)
Coating area per well: 1.90 cm²
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty