Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol, Laboratory Reagent

Supplier: 0000019890
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

M2520-1L M2520-20L M2520-4L M2520-500ML
101204-656EA 100.09 USD InStock
101204-656 101204-300 101205-024 101202-962
Methanol, Laboratory Reagent
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty