Would you like to log in again?
Antipain-Dihydrochloride
Supplier: G-Biosciences
786-045 786-045
82022-376EA 158.89 USD InStock
CA82022-376 82022-376
Antipain-Dihydrochloride
Proteins and Peptides
A selection of individual protease inhibitors are offered for researchers who wish to design their own cocktails, supplement existing cocktails, or screen for specific proteases
  • Inhibits papain, trypsin, and plasmin.
Also available is the Protease Inhibitor Set, which contains all 12 of our individual protease inhibitors, each in a ready-to-use solution at a 100X concentration. The 1X concentration of the protease inhibitors is designed to give >90% inhibition in crude tissue extracts. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity.
G-Biosciences offers the following individual protease inhibitors as a dry powder: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pepstatin, Phosphoramidon, and PMSF.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty