Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ACE2 (Human) ELISA Kit, BioVision

Supplier: BioVision
AB285246-96TEST
76236-024EA 1310.03 USD InStock
76236-024
ACE2 (Human) ELISA Kit, BioVision
Assays ELISAs
A Sandwich ELISA kit for the quantitative measurement of ACE2 in biological fluids.

  • Easy, convenient and time-saving method to measure the level of ACE2 in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0.391-25 ng/ml
  • Sensitivity: < 0.234 ng/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%
  • This assay has high sensitivity and excellent specificity for detection of ACE2. No significant cross-reactivity or interference between ACE2 and analogues was observed.

Angiotensin-converting enzyme, or ACE, is a central component of the renin-angiotensin system (RAS), which controls blood pressure by regulating the volume of fluids in the body. It converts the hormone angiotensin I to the active vasoconstrictor angiotensin II. Therefore, ACE indirectly increases blood pressure by causing blood vessels to constrict. BioVision’s ACE2 ELISA kit is a sandwich ELISA assay for the quantitative measurement of ACE2 in Human serum, plasma and cell culture supernatants. The density of color is proportional to the amount of ACE2 captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty