Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CHO Host Cell Proteins Elisa Kit

Supplier: BioVision
AB285247-96TEST
76236-040EA 1847.73 USD InStock
76236-040
CHO Host Cell Proteins Elisa Kit
Assays ELISAs
A Sandwich ELISA kit for the quantitative measurement of CHO host cell protein in biological fluids.

  • Easy, convenient and time-saving method to measure the level of CHO HCP in serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 0.1 to 100 ng/ml
  • Sensitivity: <0.31 ng/ml
  • Assay Precision: Intra-Assay: CV <8%; Inter-Assay: CV <10%
  • This assay has high sensitivity and excellent specificity for detection of CHO HCP. No significant cross-reactivity or interference between CHO HCP and analogues was observed

CHO cell line is widely used in the expression of therapeutic protein , which is a cost effective method for production of commercial quantities of a drug substance. One of the goal in designing a downstream process for biopharmaceutical products is to remove any possible contaminants, including host cell proteins. HCPs can still be present at significant concentrations following several purification steps and can even be co-purified and concentrated with the drug substance itself. HCP in the products will result in adverse toxic or immunological reactions which compromise product safety and quality. This it is desirable to reduce the host cell protein contamination to the lowest level. Therefore, during the development of a downstream process, suitable assays must be available to determine the amount of HCP present.. BioVision’s CHO HCP ELISA kit is a sandwich ELISA assay for the quantitative measurement of CHO HCP in Human serum, plasma and cell culture supernatants. The density of color is proportional to the amount of CHO HCP captured from the samples.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty