Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S-Adenosylmethionine (SAM) ELISA Kit, BioVision, Inc.

Supplier: BioVision
AB287822-96TEST
76236-050EA 1445.73 USD InStock
76236-050
S-Adenosylmethionine (SAM) ELISA Kit, BioVision, Inc.
Assays ELISAs
A competitive ELISA kit for the quantitative measurement of S-Adenosylmethionine in bio-fluids

  • Easy, convenient and time-saving method to measure the level of SAM in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 0.39 – 25 μg/ml
  • Sensitivity: < 0.234 μg/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%
  • No significant cross-reactivity or interference with its analogues was observed.

S-Adenosylmethionine (SAM) is a common cosubstrate involved in methyl group transfers, transsulfuration, and aminopropylation. Although these anabolic reactions occur throughout the body, most SAM-e is produced and consumed in the liver. In bacteria, SAM-e is bound by the SAM riboswitch, which regulates genes involved in methionine or cysteine biosynthesis. In eukaryotic cells, SAM-e serves as a regulator of a variety of processes including DNA, tRNA, and rRNA methylation; immune response; amino acid metabolism; transsulfuration; and more. In plants, SAM-e is crucial to the biosynthesis of ethylene, an important plant hormone and signaling molecule. BioVision’s SAM ELISA kit is a sandwich ELISA assay for the quantitative measurement of SAM in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of SAM captured from the samples.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty