Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CHO/K1 Whole Cell Lysate, Rockland Immunochemical

Supplier: Rockland Immunochemicals
W07-000-359
10802-374EA 162.98 USD InStock
10802-374
CHO/K1 Whole Cell Lysate, Rockland Immunochemical
Protein Extraction Protein Extraction Reagents
Ready-to-use whole cell lysates are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality, protein integrity and lot-to-lot reproducibility.

  • Synonyms: CHO/K1 Whole Cell Lysate, CHO K1 WCL, CHO-K1 lysate, CHOK1 lysate
  • Stabilizer: 10% (v/v) Glycerol
  • Origin: Chinese Hamster
  • Cell Line: Chinese Hamster Ovary (CHO)

1X SDS-PAGE Sample Buffer (62.5 mM Tris HCl, 2% SDS, 10% Glycerol and 0.005% bromophenol blue, pH 6.8) is used.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty