Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indo-1 AM calcium sensor dye

Supplier: Anaspec Inc
Synonyms: 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylesterIndo-1-(acetoxymethyl) esterIndo-1 AM Ester

AS-84006
103011-166EA 337.62 USD InStock
103011-166
Indo-1 AM calcium sensor dye
Indo-1 AM
Emission-ratiometric fluorescent indicator for quantifying intracellular Ca2+ concentration.

  • Storage: -20°C desiccated and protected from light
  • Spectral Properties: Abs/Em = 346/475 nm
  • Solvent System: DMSO


Formula: C₄₇H₅₁N₃O₂₂
MW: 1009.91 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036955
CAS Number: 112926-02-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty