Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IL-11 (Human) ELISA Kit, BioVision

Supplier: BioVision
AB285297-96TEST
76265-586EA 1383.88 USD InStock
76265-586
IL-11 (Human) ELISA Kit, BioVision
Assays ELISAs
An Sandwich ELISA kit for quantifying human IL-11 in human serum, plasma & other biological fluids.

  • Easy, convenient and time-saving method to measure the level of IL-11 in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 31.25 - 2000 pg/ml
  • Sensitivity: < 18.75 pg/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%

IL-11 is a cytokine that stimulates the proliferation of hematopoietic stem cells and megakaryocyte progenitor cells and induces megakaryocyte maturation resulting in increased platelet production. Also promotes the proliferation of hepatocytes in response to liver damage. Binding to its receptor formed by IL6ST and either IL11RA1 or IL11RA2 activates a signaling cascade that promotes cell proliferation. Signaling leads to the activation of intracellular protein kinases and the phosphorylation of STAT3. BioVision’s IL-11 ELISA kit is a sandwich ELISA assay for the quantitative measurement of human IL-9 in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human IL-11 captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty