Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amyloid Beta 42 (Human) ELISA Kit, BioVision

Supplier: BioVision
AB289832-100TEST
76265-034EA 1310.03 USD InStock
76265-034
Amyloid Beta 42 (Human) ELISA Kit, BioVision
Assays ELISAs
An Sandwich ELISA kit for quantifying human amyloid beta 42 alpha in biological fluids.

  • Easy, convenient and time-saving method to measure the level of Amyloid beta 24 alpha in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids.
  • Detection Range: 15.62 - 1000 pg/ml
  • Sensitivity: < 9.375 pg/ml
  • Assay Precision: Intra-Assay: CV < 8%; Inter-Assay: CV < 10%

Amyloid beta (Aβ or Abeta) denotes peptides of 36–43 amino acids that are crucially involved in Alzheimer's disease as the main component of the amyloid plaques found in the brains of Alzheimer patients. The peptides result from the amyloid precursor protein (APP), which is cleaved by beta secretase and gamma secretase to yield Aβ. Aβ molecules can aggregate to form flexible soluble oligomers which may exist in several forms. It is now believed that certain misfolded oligomers (known as "seeds") can induce other Aβ molecules to also take the misfolded oligomeric form, leading to a chain reaction akin to a prion infection. The seeds or the resulting amyloid plaques are toxic to nerve cells. The other protein implicated in Alzheimer's disease, tau protein, also forms such prion-like misfolded oligomers, and there is some evidence that misfolded Aβ can induce tau to misfold. BioVision’s Amyloid Beta 42 ELISA kit is a sandwich ELISA assay for the quantitative measurement of human Amyloid Beta 42 in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human Amyloid Beta 42 captured from the samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty