Would you like to log in again?
Crystal Violet Cell Cytotoxicity Assay Kit, BioVision
Supplier: BioVision
AB232855-1000TEST
76265-120EA 494.79 USD InStock
76265-120
Crystal Violet Cell Cytotoxicity Assay Kit, BioVision
Assays Cellular Assays
Kit offers an excellent method for in vitro cytotoxicity studies & high-throughput drug screening
  • Simple procedure; takes less than 2 hours
  • Fast, sensitive and convenient
Crystal violet (CV) cell cytotoxicity assay is one of the common methods used to detect cell viability or drug cytotoxicity. CV is a triarylmethane dye that can bind to ribose type molecules such as DNA in nuclei. Normally, dead adherent cells will detach from cell culture plates and will be removed from viable cell propulation during washing steps. CV staining can be used to quantify the total DNA of the remaining population and thus determine cell viability. The CV staining is directly proportional to the cell biomass and can be measured at 570 nm. CV staining is a quick and versatile assay for screening cell viability under diverse stimulation or inhibition conditions. BioVision’s CV cell cytotoxicity assay kit is simple, accurate, reproducible and sensitive. It also includes Doxorubicin as a positive control. This kit offers an excellent and efficient method for in vitro cytotoxicity studies as well as high-throughput drug screening
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty