Would you like to log in again?
Phospholipase A2 Activity Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB273278-100TEST
76265-124EA 904.55 USD InStock
76265-124
Phospholipase A2 Activity Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Highly Sensitive Assay, HTS
  • Simple, rapid & convenient
  • The assay can detect total phospholipase A2 (cytosolic and secretory). Activity less than 0.5 mUin various sample types.
Phospholipase(s) A2 (PLA2s, EC 3.1.1.4) are a family of enzymes that catalyze the hydrolysis an acyl ester bond in the sn-2 position of a glycerophospholipid to release a lysophospholipid and a free fatty acid. Members of the PLA2 superfamily can be divided into three major subtypes based upon their size and cation dependence. Two of the subtypes, the low molecular weight secretory phospholipase A2 enzymes (sPLA2) and the high molecular weight cytosolic enzymes (cPLA2), are characterized by their requirement of Ca2+ for catalytic activity. The 14 kDa sPLA2 enzymes are known to play an important role in the digestion of dietary phospholipids, host defense against bacterial infections as well as production of eicosanoids. The 85 kDa cPLA2 enzymes play an important role in inflammatory disorders by releasing arachidonic acid from membrane phospholipids for production of various eicosanoids. The third subtype, the high molecular weight calcium-independent phospholipases A2 (iPLA2), are intracellular forms of the enzyme that have been recently implicated in functions such as phospholipid remodeling, cell proliferation and fat catabolism. BioVision’s Phospholipase A2 Activity Assay Kit provides a quick, sensitive and simple way for measuring PLA2 activity in various biological samples. In this assay, active PLA2 cleaves a synthetic thiophospholipid, producing a lysothiophospholipid which reacts with a fluorogenic probe to produce a highly fluorescent product detectable in the visible range (Ex/Em= 388/513 nm). The assay is simple to perform, high-throughput adaptable and can detect less than 0.1 mU of PLA2 activity
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty