Would you like to log in again?
Gelatinase (Gelatin Degradation/Zymography) Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB234057-100TEST
76265-126EA 643.53 USD InStock
76265-126
Gelatinase (Gelatin Degradation/Zymography) Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Sensitive Fluorometric Assay to Detect Gelatinase in Mammalian Samples
  • Rapid & sensitive
  • Simple assay to measure gelatinase activity as well as to screen/study/characterize potential inhibitors of gelatinase
  • Includes Fluorometric Gelatinase Substrate and Positive Control
Gelatinases are a type of matrix zinc-dependent metalloproteases (MMPs) that degrade gelatins and a variety of other extracellular matrix proteins. These enzymes are synthesized as latent zymogens that are activated by proteolysis and inhibited by tissue inhibitors of metalloproteases (TIMPs). Two mammalian gelatinases, Gelatinase A (MMP-2) and Gelatinase B (MMP-9), are critical for basement membrane degradation and are highly upregulated in variety of tumor cells. Gelatinase activity is usually detected by small peptide-based activity assays which may suffer from lack of substrate specificity. Other methods for gelatinase activity include gelatin Zymography where samples are electrophoresed on a gelatin-containing SDS-PAGE, and further renatured in a suitable buffer for 12-16 h. The zymogram is subsequently stained, and areas of digestion appear as clear bands against a darkly stained background where the substrate has been degraded by the enzyme. Such methods are laborious, time-consuming and may lead to the loss of enzymatic activity as renaturation may not be completely reversible. BioVision’s Gelatinase Activity Assay Kit utilizes a hybrid approach for the detection of gelatinase activity by employing a highly quenched gelatin substrate which upon cleavage by a suitable gelatinase releases a fluorophore, which can be easily quantified using a conventional microplate reader. This method is substrate-specific, simple, fast, high-throughput adaptable and amenable to the sensitive detection of gelatinase activity (as low as 0.06 mCDU for bacterial collagenase) in biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty