Would you like to log in again?
HDAC6 Activity Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB284549-100TEST
76265-266EA 956.9 USD InStock
76265-266
HDAC6 Activity Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
High-throughput compatible, quick & easy screening for HDAC6 activity in biological samples.
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of HDAC6 inhibitors
  • Kit includes Tubacin, an HDAC6 inhibitor, and a stable, recombinant human HDAC6
Histone Deacetylase 6 (HDAC6, EC 3.5.1.98), is a class IIb HDAC enzyme that deacetylates an ε-N-acetyl lysine of histone and non-histone protein substrates. It is a unique member of HDAC family as it contains two deacetylase domains that are proposed to function independent of each other. HDAC6 can shuttle between the nucleus and cytoplasm, suggesting potential extra-nuclear functions by regulating the acetylation status of non-histone substrates. HDAC6 also affects transcription and translation by regulating the heat-shock protein 90 (Hsp90) and stress granules. Elevated HDAC6 activity is associated with cell motility and increases α-tubulin deacetylation, thus influencing cancer cell metastasis. In addition, mutations in HDAC6 gene have been associated with Alzheimer's disease. HDAC6-selective inhibitors are considered as promising targets for autoimmune, oncology and inflammatory diseases. BioVision’s HDAC6 Activity Assay Kit utilizes deacetylase activity of HDAC6 towards a synthetic acetylated-peptide substrate resulting in the release of an AFC fluorophore, which can be easily quantified using a conventional microplate reader. This simple and high-throughput adaptable assay kit can be used to detect HDAC6 activity (as low as 3 U) in complex biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty