Would you like to log in again?
Collagenase (Collagen Degradation/Zymography) Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB234624-100TEST
76265-130EA 645.13 USD InStock
76265-130
Collagenase (Collagen Degradation/Zymography) Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Sensitive Fluorometric Assay to Detect Collagenase in Mammalian Samples
  • Rapid & sensitive
  • Simple assay to measure collagenase activity as well as to screen/study/characterize potential inhibitors of collagenase
  • Includes Fluorometric Collagenase Substrate and Positive Control
Collagenases, members of the matrix metalloproteinase (MMP) family, are endopeptidases that digest native collagen in the triple helix region and are involved in the physiological and pathological turnover of connective tissues. Collagens are the major fibrous component of animal extracellular connective tissue. Bacterial collagenases differ from vertebrate collagenases in that they exhibit broader substrate specificity. Collagenases may play a role in facilitating tumor cell invasion of the extracellular matrix at multiple stages of the metastatic process. Collagenases have also been approved for medical uses for the treatment of Dupuytren's contracture, Peyronie's disease and wound healing. Collagenase activity is usually detected by small peptide-based activity assays which may suffer from lack of substrate specificity. Other methods for collagenase activity include collagen Zymography where samples are electrophoresed on a collagen-containing SDS-PAGE, and further renatured in a suitable buffer for 12-16 h. The zymogram is subsequently stained, and areas of digestion appear as clear bands against a darkly stained background where the substrate has been degraded by the enzyme. Such methods are laborious, time-consuming and may lead to the loss of enzymatic activity as renaturation may not be completely reversible. BioVision’s Collagenase Activity Assay Kit utilizes a hybrid approach for the detection of collagenase activity by employing a highly quenched collagen substrate which upon cleavage by a suitable collagenase releases a fluorophore, which can be easily quantified using a conventional microplate reader. This method is substrate-specific, simple, fast, high-throughput adaptable and amenable to the sensitive detection of collagenase activity (as low as 0.6 mCDU for bacterial collagenase) in biological samples
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty