Would you like to log in again?
IDH1 R132H Mutant Inhibitor Screening Kit (Colorimetric), BioVision, Inc.
Supplier: BioVision
AB283394-100TEST
76264-664EA 1243.27 USD InStock
76264-664
IDH1 R132H Mutant Inhibitor Screening Kit (Colorimetric), BioVision, Inc.
Assays Enzyme Assays
This assay has been designed for screen/study/characterizing potential IDH1 R132H Mutant inhibitors
  • Simple procedure
  • Fast and convenient
  • Screening/studying/characterizing potential Mutant IDH inhibitors
In eukaryotic cells, Isocitrate Dehydrogenase (IDH1, IDH2 and IDH3) is an enzyme that catalyzes the decarboxylation of Isocitrate producing α-Ketoglutarate and CO2. Mutations of the different isoforms of IDH, IDH1 and IDH2, are commonly found in human cancers. Among these mutants, the cytosolic IDH1 mutant at the Arginine 132 position substitution with Histidine (IDH1 R132H) is the most commonly mutation found in primary human brain cancers. Isocitrate Dehydrogenase Mutant (IDH mutant) causes a “gain-of-function” mutation, which reduces its affinity for isocitrate and favors the conversion of α-ketoglutarate to D-2-Hydroxyglutarate (D2HG). High level of D2HG correlates with an increased risk for malignant brain tumors. Thus, the search for potential novel as well as specific inhibitors for this target has increased considerably in recent years. In BioVision’s IDH1 R132H Mutant Inhibitor Screening Kit, IDH1 R132H Mutant oxidizes NADPH into NADP+, which decreases the absorbance at 340 nm. In the presence of IDH1 R132H Mutant inhibitor, the reaction is impeded. An IDH1 R132H Mutant Inhibitor Control is included to compare the efficacy of the sample inhibitors. The assay is high-throughput adaptable and can be completed in less than 1 hr.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty