Would you like to log in again?
Diamine Oxidase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB241004-100TEST
76264-666EA 959.34 USD InStock
76264-666
Diamine Oxidase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
Simple, rapid & convenient assay to measure Diamine Oxidase Activity. Detection limit: 1 µU.
  • Rapid & Convenient
  • The assay can detect as little as 1 µU of DAO enzymatic activity.
  • Includes DAO Enzyme as Positive Control
I. Diamine Oxidase (DAO, E.C. 1.4.3.22) is an important regulator of polyamine levels in human physiology. DAO metabolizes the oxidative conversion of biogenic polyamines such as putrescine and cadaverine into aldehydes. Biogenic amines are known regulators of important biological pathways such as cell growth and signal transduction, placing DAO in an important regulatory position in maintaining cell viability. Putrescine itself is also an activator of c-jun and c-fos expression, further linking this diamine with potential roles in cancer development. DAO is notably involved in regulating growth and differentiation of intestinal mucosa, bone marrow, and other rapidly proliferating tissue. In addition to its role in tissue growth, DAO activity is important in metabolism. Loss of diamine oxidase activity can lead to histamine intolerance and histamine poisoning. BioVision’s Diamine Oxidase Activity Assay Kit provides a straightforward method to determine DAO activity of tissue lysates as well as recombinant enzymes, with a detection limit of less than 1 pmole/min of activity. In the assay, DAO converts the provided substrate, yielding an intermediate and H2¬¬O2. H2¬O2 is then utilized by the DAO Enzyme Mix to generate fluorescence (Ex/Em = 535/587 nm) from the DAO Probe.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty