Would you like to log in again?
EZCell™ Direct Glucose Uptake Assay Kit, BioVision
Supplier: BioVision
AB234043-50TEST
76265-132EA 1213.82 USD InStock
76265-132
EZCell™ Direct Glucose Uptake Assay Kit, BioVision
Assays Cellular Assays
Non-radioactive, Highly Sensitive Assay
  • Simple procedure
  • Fast and convenient
  • The assay is easy, non-radioactive, and highly sensitive
Glucose uptake is one of the key processes for cellular glucose metabolism. The study of glucose uptake can provide important information for understanding glucose metabolism and regulation in normal and disease development such as diabetes. BioVision’s EZCellTM Glucose Uptake Assay kit is simple, ultra-sensitive and easy to use. A specific hexokinase inhibitor that inhibits hexokinase, the first enzyme metabolizing glucose in cells is used to arrest glucose consumption after its uptake. Glucose Uptake is measured by using a set of enzymatic reactions that specifically oxidize glucose producing intermediates that react with the OxiRed™ Probe generating a fluorescence signal (Ex/Em=535/587 nm). The fluorescence signal is directly proportional to the amount of glucose that has been taken up and accumulated inside the cells. Unlike other kits detecting glucose derivatives, this glucose uptake assay provides a direct, powerful tool for studying this process as well as for screening and characterization of drugs that regulate glucose uptake during normal and disease development
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty