Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRA5 Mouse Monoclonal Antibody

Supplier: BioVision
AB286170-50UG
76264-734EA 1057.21 USD InStock
76264-734
Anti-GRA5 Mouse Monoclonal Antibody
Antibodies
Antibody directed against the GRA5 protein from the intracellular protozoan parasite Toxoplasma gondii. The protein is distributed within the dense granules secretory organelles and delineates the parasitophorous vacuolar membrane. The protein also localizes in the cyst wall.

Type: Primary
Antigen: GRA5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Toxoplasma gondii

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty