Would you like to log in again?
MASTALEX™ MRSA Latex Kit, Hardy Diagnostics
Supplier: Hardy Diagnostics
RST501
76220-914EA 601.02 USD InStock
76220-914
MASTALEX™ MRSA Latex Kit, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
MASTALEX™ Kit detects Penicillin Binding Protein 2'(PBP2') in isolates of Staphylococcus species. This assists in the identification of Methicillin Resistant Staphylococcus aureus and Methicillin Resistant coagulase negative Staphylococci.
  • Convenient latex format
  • Clear and easy to read
  • Rapid turn round time
  • Results available in 5 to 10 minutes
  • Sensitive, user friendly and cost effective
  • May be used for screening or diagnosis
  • Color coded reagents
PBP2' is coded by the mecA gene and is present in the membranes of Methicillin Resistant Staphylococcus species. The latex particles are coated with antibodies specific for the PBP2' protein. The kit uses monoclonal antibody directed against PBP2'.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty