Would you like to log in again?
Buffered Charcoal Yeast Extract (BCYE) with DGVP, Hardy Diagnostics
Supplier: Hardy Diagnostics
G209
76213-942PK 84.71 USD InStock
76213-942
Buffered Charcoal Yeast Extract (BCYE) with DGVP, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Buffered Charcoal Yeast Extract (BCYE) Agar formulations are recommended for use in the cultivation and primary isolation of Legionella species. in water and other samples suspected of harboring the bacteria.
  • Optically clear polystyrene for distortion free viewing
  • Heavy-duty reduces breakage during shipping
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
  • Packaged in sleeves of 10 plates in a "breathable" bag that prevents buildup of condensation and excess moisture
BCYE with DGVP contains the dyes bromcresol purple and bromothymol blue, along with glycine, vancomycin and polymyxin. Antibiotics in the medium improve the recovery of Legionella species by inhibiting the growth of contaminating microorganisms, while dyes in the medium facilitate the differentiation and identification of Legionella.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty