Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Poly-L-lysine hydrobromide

Supplier: MP Biomedicals
0219454405
76204-134EA 92.96 USD InStock
76204-134
Poly-L-lysine hydrobromide
Proteins and Peptides
Poly-L-lysine is a positively charged amino acid polymer. Poly-lysine binds to DNA, red cell membrane and any negatively charged protein. It is typically used as a coating substrate for culture dishes, slides, etc. It enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When adsorbed to the culture surface, poly-lysine increases the number of positively charged sites available for cell binding.

  • Grade : Cell Culture Reagent, γ-irradiated
  • Suitability : suitable for cell culture
  • Key Applications : Attachment factor for tissue culture.
  • Application Areas : Cell Biology & Analysis
  • Product Type : Biochemicals
  • Presentation : Beige Powder
  • NOTES : There is approximately one HBr or HCl per lysine unit. The HBr/HCl allows it to be a crystalline solid and soluble in water. To remove the HBr/HCl, solubilize the product in a neutral buffer and dialyze away the salts.
  • Solubility : Soluble in water (50mg/mL) clear,colorless solution.
  • Sterility : γ-Irradiated

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty