Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium acetate xhydrate 99.0-100.5% (by anhydrous basis), Puriss., meets analytical specification of FCC, E263, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Calcium acetate hydrateAcetic acid calcium salt hydrateCalcium di(acetate) hydrate

25011-6X1KG
BJ25011-1KGCS 1339.13 USD InStock
BJ25011-1KG
Calcium acetate xhydrate 99.0-100.5% (by anhydrous basis), Puriss., meets analytical specification of FCC, E263, Fluka™
Calcium acetate xhydrate


Formula: (H₃CCOO)₂Ca·nH₂O
MW: 176.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150019
CAS Number: 114460-21-8
Order Now
Specification Test Results
Assay (calculated on the anhydrous basis) 99.0 - 100.5 %
Water (Karl Fischer) Max. 7.0 %
Water insoluble matter Max. 0.3 %
pH (10 %, 20°C) 7 - 9.0
Arsenic (As) Max. 1 ppm
Copper (Cu) Max. 0.001 %
Iron (Fe) Max. 0.002 %
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.03 %
Fluoride (F) Max. 0.005 %
Sulfate (SO₄) Max. 0.05 %
Formic acid, formiates other impurities (HCOOH) Max. 1000 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty