Would you like to log in again?
Plastic Test Tube Rack, 12 Tubes, United Scientific Supplies
Supplier: United Scientific Supplies
TTHP03-VWR
76263-164EA 5.86 USD InStock
76263-164
Plastic Test Tube Rack, 12 Tubes, United Scientific Supplies
Racks Tube Racks
Store and display your hydrometers.
  • Easy to assemble
  • Chemical resistant
Sturdy polypropylene rack will hold 12 test tubes up to 20 mm in diameter. Easy to assemble, and chemical resistant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty