Would you like to log in again?
Flask Stand, United Scientific Supplies
Supplier: United Scientific Supplies
45105-VWR
76263-204EA 9.75 USD InStock
76263-204
Flask Stand, United Scientific Supplies
Supports Flask Supports
Designed for use with flasks up to 10 l, this polypropylene flask stand is ribbed to safely hold flasks in place.
  • Autoclavable
This non-corrosive separatory funnel holder, molded in polypropylene, has a front opening which provides clear view of the solution and allows unobstructed placement of separating funnels. Two tapered wedges are provided to ensure firm grip on rods measuring 1/2" or 3/8" diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty