Would you like to log in again?
Plastic Test Tube Rack Cube, United Scientific Supplies
Supplier: United Scientific Supplies
76001
76263-262EA 31.93 USD InStock
76263-262
Plastic Test Tube Rack Cube, United Scientific Supplies
Racks Tube Racks
Modular design with various sizes.
  • Multiple sizes
  • Modular design
A very sturdy, one-piece, molded polypropylene rack. All four sides can be used interchangeable for 12 mm through 25 mm diameter tubes. Rack features a total of 88 spaces on its four sides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty