Would you like to log in again?
Test Tube Rack with Glass Tubes, United Scientific Supplies
Supplier: United Scientific Supplies
TTRSET-VWR
76263-266EA 22.25 USD InStock
76263-266
Test Tube Rack with Glass Tubes, United Scientific Supplies
Racks Tube Racks
Ideal for collecting outdoor samples.
  • Two-tiered tube rack
  • Holds (12) 24 ml glass tubes (included)
Conical glass tubes have tight fitting rubber stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty