Would you like to log in again?
Plastic Test Tube Rack, 6 Place, United Scientific Supplies
Supplier: United Scientific Supplies
TTHP01
76263-268EA 4.68 USD InStock
76263-268
Plastic Test Tube Rack, 6 Place, United Scientific Supplies
Racks Tube Racks
Economical yet very durable rack.
  • Easy to clean
  • Polyethylene
This polyethylene rack holds six test tubes, and has six drying pins. Sides are fully open for clear viewing. Rack has two 30 mm and four 20 mm holes, and is supplied pre-assembled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty