Would you like to log in again?
Plastic Test Tube Rack, 6 Places, Unassembled, United Scientific Supplies
Supplier: United Scientific Supplies
TTHP02
76263-270EA 2.47 USD InStock
76263-270
Plastic Test Tube Rack, 6 Places, Unassembled, United Scientific Supplies
Racks Tube Racks
Economical yet very durable rack.
  • Easy to clean
  • Polyethylene
This polyethylene rack will hold six test tubes, and has six drying pins. Sides are fully open for clear viewing. Rack has six 25 mm holes and requires assembly.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty