Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene-α-sulfonyl fluoride ≥99.0%, white powder, Proteomics Grade

Supplier: VWR
Danger
Synonyms: α-Toluenesulfonyl fluorideα-Toluenesulphonyl fluoridePMSFPhenylmethylsulfonyl fluoridePhenylmethylsulphonyl fluoridePhenylmethanesulfonyl fluoridePhenylmethanesulphonyl fluoride

M145-25G M145-5G
97064-898EA 229.52 USD InStock
97064-898 97064-900
Toluene-α-sulfonyl fluoride ≥99.0%, white powder, Proteomics Grade
Toluene-α-sulphonyl fluoride


Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
UN: 2928
ADR: 6.1,II
Order Now
Specification Test Results
Identification PASS / Fail
Melting Point 91 - 95 °C
Purity ≥99.0%
Solubility (5%, EtOH) PASS / Fail
Thin Layer Chromatography (TLC) ONE SPOT / Fail

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty