Would you like to log in again?
QuantiChrom™ Indole Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DIND-100
76237-108EA 283.62 USD InStock
76237-108
QuantiChrom™ Indole Assay Kit, BioAssay Systems
Assays Cellular Assays
Indole determination in biological samples.
  • Fast and sensitive, convenient
  • Use of 100µl sample
  • Linear detection range from 3 to 100 µM indole in 96-well plate assay
  • The procedure involves adding a single working reagent, and reading the absorbance immediately
  • Method: OD565 nm
  • Samples: Bacterial growth medium
Indole is the primary product of tryptophan breakdown by tryptophanase. The indole test is commonly performed on bacteria to classify them on their ability to break down tryptophan to indole. Indole assay kit is based on a modified version of Ehlrich's and Kovac's reagents, which reacts with indole to produce a colored compound at 565 nm. The intensity of this colored compound is directly proportional to the indole in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty