Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EnzyChrom™ Acetaldehyde Assay Kit, BioAssay Systems

Supplier: BioAssay Systems
EACT-100
76236-116EA 476.4 USD InStock
76236-116
EnzyChrom™ Acetaldehyde Assay Kit, BioAssay Systems
Assays Cellular Assays
Acetaldehyde determination in biological and food or beverage samples.

  • Fast and sensitive
  • Linear detection range (10 µl sample): 4 µM to 4 mM acetaldehyde in 96-well plate assay
  • High-throughput
  • Room temperature assay, no 37 °C heater is needed, convenient
  • Homogeneous "mix-incubate-measure" type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: OD565 nm

The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Acetaldehyde (CH₃CHO) is one of the most widely occurring aldehydes in nature and is commonly used in industry. Acetaldehyde, a metabolic byproduct of ethanol in the liver, is toxic to the human body and is rapidly converted to the less harmful acetic acid by the enzyme aldehyde dehydrogenase. People with a deficiency of aldehyde dehydrogenase accumulate acetaldehyde when consuming alcohol and this accumulation results in facial and body flushing often referred to as "Asian flush syndrome". Build up of acetaldehyde has also been associated with the effects of hangovers from alcohol consumption. Although classified as a carcinogen, acetaldehyde is naturally found in many foods and beverages such as ripe fruit, coffee, and wine. BioAssay Systems colorimetric acetaldehyde assay kit is based on aldehyde dehydrogenase catalyzed oxidation of acetaldehyde, in which the formed NADH reduces a formazan reagent. The intensity of the product color, measured at 565 nm, is directly proportional to the acetaldehyde concentration in the sample.

Samples: Biological samples (example: plasma, serum, urine, tissue and culture media) and food or beverage samples (example: wine, coffee, and juice).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty